• USA Home
  • Search Results

  • larginine174207479311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Arginine

10 Products
Synonym:   (S)-2-Amino-5-guanidinopentanoic acid
Image of <SC>L</SC>-Arginine
CAS Number:
Linear Formula:
 H2NC(=NH)NH(CH2)3CH(NH2)CO2H
Molecular Weight:
 174.20
 Beilstein Registry Number:
 1725413
 EC Number:
 FEMA Number:
 3819
 Flavis number:
 17.003
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  UV-Absorption Values
  Suitability
  Product Line
  Solubility
  Biological Source
  Special Grade
  Physical Form
  Assay
Show More