• USA Home
  • Search Results

  • largininemonohydrochloride21066111934211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Arginine monohydrochloride

7 Products
Synonym:   (S)-(+)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid monohydrochloride, (S)-(+)-Arginine hydrochloride
Image of <SC>L</SC>-Arginine monohydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C6H14N4O2 · HCl
Molecular Weight:
 210.66
 Beilstein/REAXYS Number:
 3631658
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Restriction
  Vapor pressure
  UV-Absorption Values
  Potency
  Bulk Density
  pH Value
  Product Line
  Special Grade
  Color
  Application(s)
  Density
  Solubility
  Physical Form
  Melting Point
  Assay
Show More