• USA Home
  • Search Results

  • lasparagine132127047311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Asparagine

4 Products
Synonym:   (S)-2-Aminosuccinic acid 4-amide, L-Aspartic acid 4-amide
Image of <SC>L</SC>-Asparagine
CAS Number:
Linear Formula:
 H2NCOCH2CH(NH2)CO2H
Molecular Weight:
 132.12
 Beilstein/REAXYS Number:
 1723527
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
  Product Line
  Color
  Physical Form
  Assay
Show More