• USA Home
  • Search Results

  • lasparticacid133105684811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Aspartic acid

10 Products
Synonym:   (S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid
Image of <SC>L</SC>-Aspartic acid
CAS Number:
Linear Formula:
 HO2CCH2CH(NH2)CO2H
Molecular Weight:
 133.10
 Beilstein/REAXYS Number:
 1723530
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  UV-Absorption Values
  Manufacturer designation
  Agency/Method
  Biological Source
  Product Line
  Color
  Special Grade
  Application(s)
  Physical Form
  Solubility
Show More