• USA Home
  • Search Results

  • lasparticacid13c415n1380720246827711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Aspartic acid-13C4,15N

1 Products
Image of <SC>L</SC>-Aspartic acid-<SUP>13</SUP>C<SUB>4</SUB>,<SUP>15</SUP>N
CAS Number:
Linear Formula:
 HO213C13CH213CH(15NH2)13CO2H
Molecular Weight:
 138.07
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
Show More