• USA Home
  • Search Results

  • lasparticacidpotassiumsalt17119111563511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Aspartic acid potassium salt

1 Products
Synonym:   Potassium L-aspartate
Image of <SC>L</SC>-Aspartic acid potassium salt
CAS Number:
Linear Formula:
 HO2CCH2CH(NH2)CO2K
Molecular Weight:
 171.19
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Color
Show More