• USA Home
  • Search Results

  • latanoprost4325913020982411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Latanoprost

4 Products
Synonym:   Isopropyl (5Z,9α,11α,15R)-9,11,15-trihydroxy-17-phenyl-18,19,20-trinor-prost-5-en-1-oate, Isopropyl (Z)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate, Xalatan
Image of Latanoprost
CAS Number:
Empirical Formula (Hill Notation):
 C26H40O5
Molecular Weight:
 432.59
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Solubility
  Manufacturer designation
Show More