• USA Home
  • Search Results

  • laurylmethacrylate2544114290511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lauryl methacrylate

1 Products
Synonym:   Dodecyl methacrylate
Image of Lauryl methacrylate
CAS Number:
Linear Formula:
 CH2=C(CH3)COO(CH2)11CH3
Molecular Weight:
 254.41
 Beilstein/REAXYS Number:
 1708160
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More