• USA Home
  • Search Results

  • lavenderoil12345800028011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lavender oil

2 Products
CAS Number:
 MDL number:
 NACRES
 NA.22
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
  Biological Source
Show More