• USA Home
  • Search Results

  • lcarnitinehydrochloride19766664546111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Carnitine hydrochloride

2 Products
Synonym:   (−)-β-Hydroxy-γ-(trimethylammonio)butyrate, Vitamin BT
Image of <SC>L</SC>-Carnitine hydrochloride
CAS Number:
Linear Formula:
 (CH3)3N+CH2CH(OH)CH2CO2H · Cl-
Molecular Weight:
 197.66
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Biological Source
Show More