• USA Home
  • Search Results

  • lcarvone15022648540111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Carvone

2 Products
Synonym:   (−)-Carvone, (R)-(−)-Carvone, (R)-5-Isopropenyl-2-methyl-2-cyclohexenone, p-Mentha-6,8-dien-2-one, Carvol
Image of <SC>L</SC>-Carvone
CAS Number:
Empirical Formula (Hill Notation):
 C10H14O
Molecular Weight:
 150.22
 Beilstein/REAXYS Number:
 2206714
 EC Number:
 FEMA Number:
 2249
 Flavis number:
 7.147
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Chemical Composition
  Assay
  Agency/Method
Show More