• USA Home
  • Search Results

  • lcysteine121165290411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Cysteine

9 Products
Synonym:   (R)-2-Amino-3-mercaptopropionic acid
Image of <SC>L</SC>-Cysteine
CAS Number:
Linear Formula:
 HSCH2CH(NH2)CO2H
Molecular Weight:
 121.16
 Beilstein/REAXYS Number:
 1721408
 EC Number:
 FEMA Number:
 3263
 Flavis number:
 17.033
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  pH Value
  Potency
  Melting Point
  Loss
  Bulk Density
  Biological Source
  Autoignition temp
  Special Grade
  Analyte Suitability
  UV-Absorption Values
  Product Line
  Titertest
  Solubility
  Assay
Show More