• USA Home
  • Search Results

  • lcysteinehydrochloridemonohydrate17563704804611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Cysteine hydrochloride monohydrate

8 Products
Image of <SC>L</SC>-Cysteine hydrochloride monohydrate
CAS Number:
Linear Formula:
 HSCH2CH(NH2)COOH · HCl · H2O
Molecular Weight:
 175.63
 Beilstein/REAXYS Number:
 5158059
 EC Number:
 MDL number:
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Document Links
  Allergen
  Organoleptic property
  UV-Absorption Values
  Product Line
  Solubility
  Loss
  Biological Source
  Suitability
  Special Grade
Show More

Product #
Description
C6852 L-Cysteine hydrochloride monohydrate, from non-animal source, cell culture tested, meets EP, USP testing specifications,
C7880 L-Cysteine hydrochloride monohydrate, reagent grade, ≥98% (TLC),
C5610 L-Cysteine hydrochloride monohydrate, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture,
30129 L-Cysteine hydrochloride monohydrate, BioUltra, ≥99.0% (RT),
30122 L-Cysteine hydrochloride monohydrate, tested according to Ph Eur,
W779989 L-Cysteine hydrochloride monohydrate, FG, produced by Wacker Chemie AG, Burghausen, Germany, Food, 20 mesh,
PHR1102 L-Cysteine hydrochloride monohydrate, Pharmaceutical Secondary Standard; Certified Reference Material,
778451 L-Cysteine hydrochloride monohydrate, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.5-101.0% (Ph.Eur (2.2.20)),