• USA Home
  • Search Results

  • lcystine240305689311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Cystine

10 Products
Synonym:   (R,R)-3,3′-Dithiobis(2-aminopropionicacid)
Image of <SC>L</SC>-Cystine
CAS Number:
Linear Formula:
 [-SCH2CH(NH2)CO2H]2
Molecular Weight:
 240.30
 Beilstein/REAXYS Number:
 1728094
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Suitability
  Melting Point
  Density
  Bulk Density
  Agency/Method
  Product Line
  Loss
  Biological Source
  Color
  Special Grade
  Physical Form
  Solubility
  Assay
Show More

Product #
Description
C8755 L-Cystine, ≥98% (TLC), crystalline,
C7602 L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%,
C5735 L-Cystine, PharmaGrade, Ajinomoto, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture,
30200 L-Cystine, ≥99.7% (TLC),
49603 L-Cystine, certified reference material, TraceCERT®,
779539 L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%,
PHR1323 L-Cystine, Pharmaceutical Secondary Standard; Certified Reference Material,
1.02836 L-Cystine, suitable for biopharmaceutical production FCC,
1.02837 L-Cystine, for biochemistry,
RES1520C-A7 L-Cystine, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production., suitable for cell culture,