• USA Home
  • Search Results

  • leadiiacetatetrihydrate37933608056411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lead(II) acetate trihydrate

5 Products
Synonym:   Lead diacetate trihydrate
Image of Lead(II) acetate trihydrate
CAS Number:
Linear Formula:
 Pb(CH3CO2)2 · 3H2O
Molecular Weight:
 379.33
 Beilstein/REAXYS Number:
 3730298
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Purity Grade
  Agency/Method
  Reaction Suitability
  Special Grade
Show More