• USA Home
  • Search Results

  • lemonoil12345800856811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lemon oil

2 Products
CAS Number:
 FEMA Number:
 2625
 MDL number:
 NACRES:
 NA.21
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Chemical Composition
  Density
  Biological Source
  Document Links
  Allergen
  Organoleptic property
  Geographic origin
  Special Grade
Show More