• USA Home
  • Search Results

  • leptophos412072160990511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Leptophos

2 Products
Synonym:   O-(4-Bromo-2,5-dichlorophenyl) O-methyl phenylphosphonothioate
Image of Leptophos
CAS Number:
Empirical Formula (Hill Notation):
 C13H10BrCl2O2PS
Molecular Weight:
 412.07
 Beilstein/REAXYS Number:
 2152663
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Shelf Life
  Physical Form
  Special Grade
Show More