• USA Home
  • Search Results

  • letrozole2853011280951511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Letrozole

4 Products
Synonym:   4,4′-(1H-1,2,4-Triazol-1-ylmethylene)bisbenzonitrile
Image of Letrozole
CAS Number:
Empirical Formula (Hill Notation):
 C17H11N5
Molecular Weight:
 285.30
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
Show More