• USA Home
  • Search Results

  • leualahydrate20225anhydrousbasis7298842anhydrous11

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Leu-Ala hydrate

1 Products
Synonym:   L-Leucyl-L-alanine hydrate
Image of Leu-Ala hydrate
CAS Number:
Linear Formula:
 (CH3)2CHCH2CH(NH2)CONHCH(CH3)COOH · xH2O
Molecular Weight:
 202.25 (anhydrous basis)
 Beilstein/REAXYS Number:
 1726165
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More