• USA Home
  • Search Results

  • leucineaminopeptidasemicrosomalfromporcinekidney12345905463111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Leucine Aminopeptidase, microsomal from porcine kidney

3 Products
Synonym:   Aminopeptidase M, Aminopeptidase N, Cathepsin III, LAP, Leucinamide aminopeptidase, Leucinaminopeptidase
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.4.11.2  ( BRENDA | IUBMB )
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Type
Show More