• USA Home
  • Search Results

  • leukotrieneb4336477116024211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Leukotriene B4

1 Products
Synonym:   [5S,12R]-Dihydroxy-[6Z,8E,10E,14Z]-eicosatetraenoic acid
Image of Leukotriene B<SUB>4</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C20H32O4
Molecular Weight:
 336.47
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More