• USA Home
  • Search Results

  • leupeptin4755910347689711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Leupeptin

5 Products
Synonym:   Acetyl-Leu-Leu-Arg-al, Leupeptin hemisulfate salt, N-Acetyl-L-leucyl-L-leucyl-L-argininal hemisulfate salt
Image of Leupeptin
CAS Number:
Linear Formula:
 C20H38N6O4 · 1/2H2SO4
Molecular Weight:
 475.59
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Package quantity
  Assay
  Physical Form
Show More