• USA Home
  • Search Results

  • leupeptinhydrochloride463012412516411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Leupeptin hydrochloride

2 Products
Synonym:   Acetyl-Leu-Leu-Arg-al hydrochloride
Image of Leupeptin hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C20H38N6O4 · HCl
Molecular Weight:
 463.01
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Solubility
  Biological Source
Show More