• USA Home
  • Search Results

  • levulinicacid1161212376211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Levulinic acid

5 Products
Synonym:   4-Oxopentanoic acid, 4-Oxovaleric acid
Image of Levulinic acid
CAS Number:
Linear Formula:
 CH3COCH2CH2COOH
Molecular Weight:
 116.12
 Beilstein/REAXYS Number:
 506796
 Council of Europe no.:
 23
 EC Number:
 FEMA Number:
 2627
 Flavis number:
 8.023
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Potency
  Special Grade
  Assay
Show More