• USA Home
  • Search Results

  • lglutamicacid147135686011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Glutamic acid

10 Products
Synonym:   (S)-2-Aminopentanedioic acid, Glu
Image of <SC>L</SC>-Glutamic acid
CAS Number:
Linear Formula:
 HO2CCH2CH2CH(NH2)CO2H
Molecular Weight:
 147.13
 Beilstein/REAXYS Number:
 1723801
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Allergen
  Organoleptic property
  pH Value
  UV-Absorption Values
  Potency
  Melting Point
  Bulk Density
  Anal. Standard Form
  Manufacturer designation
  Color
  Biological Source
  Product Line
  Agency/Method
  Solubility
  Special Grade
  Application(s)
  Density
  Assay
Show More