• USA Home
  • Search Results

  • lglutamicaciddiethylesterhydrochloride23970111889411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Glutamic acid diethyl ester hydrochloride

1 Products
Image of <SC>L</SC>-Glutamic acid diethyl ester hydrochloride
CAS Number:
Linear Formula:
 C2H5OCOCH2CH2CH(NH2)COOC2H5 · HCl
Molecular Weight:
 239.70
 Beilstein/REAXYS Number:
 3597595
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More