• USA Home
  • Search Results

  • lglutamicacidg4methoxybnaphthylamide302332472350011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Glutamic acid γ-(4-methoxy-β-naphthylamide)

1 Products
Image of <SC>L</SC>-Glutamic acid γ-(4-methoxy-β-naphthylamide)
CAS Number:
Empirical Formula (Hill Notation):
 C16H18N2O4
Molecular Weight:
 302.33
 MDL number:
 NACRES:
 NA.83
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More