• USA Home
  • Search Results

  • lglutamicacidganaphthylamidemonohydrate123458101291111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Glutamic acid γ-(α-naphthylamide) monohydrate

1 Products
Image of <SC>L</SC>-Glutamic acid γ-(α-naphthylamide) monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C15H16N2O3 H2O
 MDL number:
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More