• USA Home
  • Search Results

  • lglutamicacidmonosodiumsalthydrate16911anhydrousbasis142472anhydrous11

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Glutamic acid monosodium salt hydrate

3 Products
Synonym:   (S)-2-Aminopentanedioic acid, L-Glutamic acid monosodium salt, Glu, MSG, Monosodium glutamate
Image of <SC>L</SC>-Glutamic acid monosodium salt hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C5H8NNaO4 · xH2O
Molecular Weight:
 169.11 (anhydrous basis)
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More