• USA Home
  • Search Results

  • lglutamicdehydrogenasefrombovineliver12345902912311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Glutamic Dehydrogenase from bovine liver

3 Products
Synonym:   L-GLDH, L-Glutamate:NAD[P]+ Oxidoreductase (deaminating), Glutamate Dehydrogenase from bovine liver
CAS Number:
 Enzyme Commission (EC) Number:
 1.4.1.3  ( BRENDA | IUBMB )
 MDL number:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Type
  Physical Form
Show More