• USA Home
  • Search Results

  • lglutamine146145685911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Glutamine

11 Products
Synonym:   (S)-2,5-Diamino-5-oxopentanoic acid, L-Glutamic acid 5-amide, Levoglutamide
Image of <SC>L</SC>-Glutamine
CAS Number:
Linear Formula:
 H2NCOCH2CH2CH(NH2)CO2H
Molecular Weight:
 146.14
 Beilstein/REAXYS Number:
 1723797
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  UV-Absorption Values
  Potency
  Manufacturer designation
  Density
  Bulk Density
  pH Value
  Biological Source
  Agency/Method
  Special Grade
  Color
  Product Line
  Assay
  Physical Form
  Solubility
Show More