• USA Home
  • Search Results

  • lglutathioneoxidized612632702541811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Glutathione oxidized

4 Products
Synonym:   GSSG, Glutathiol
Image of <SC>L</SC>-Glutathione oxidized
CAS Number:
Empirical Formula (Hill Notation):
 C20H32N6O12S2
Molecular Weight:
 612.63
 Beilstein/REAXYS Number:
 1718700
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Product Line
  Color
  Physical Form
  Solubility
Show More