• USA Home
  • Search Results

  • lhistidine155157100111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Histidine

9 Products
Synonym:   (S)-2-Amino-3-(4-imidazolyl)propionic acid, NSC 137773
Image of <SC>L</SC>-Histidine
CAS Number:
Empirical Formula (Hill Notation):
 C6H9N3O2
Molecular Weight:
 155.15
 Beilstein/REAXYS Number:
 84088
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 32160406
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  UV-Absorption Values
  Product Line
  Potency
  Bulk Density
  Biological Source
  Analyte Suitability
  pH Value
  Melting Point
  Physical Form
  Titertest
  Solubility
  Assay
Show More