• USA Home
  • Search Results

  • lhomoargininehydrochloride22469148301811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

L-Homoarginine hydrochloride

1 Products
Synonym:   (S)-2-Amino-6-guanidinohexanoic acid hydrochloride
Image of <SC>L</SC>-Homoarginine hydrochloride
CAS Number:
Linear Formula:
 H2NC(=NH)NH(CH2)4CH(NH2)CO2H · HCl
Molecular Weight:
 224.69
 Beilstein/REAXYS Number:
 4016621
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More