• USA Home
  • Search Results

  • licochalconea338405874922711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Licochalcone A

2 Products
Synonym:   (E)-3-[5-(1,1-Dimethyl-2-propenyl)-4-hydroxy-2-methoxyphenyl]-1-(4-hydroxyphenyl)-2-propen-1-one, 4′,4-Dihydroxy-3-α,α-dimethylallyl-6-methoxychalcone
Image of Licochalcone A
CAS Number:
Empirical Formula (Hill Notation):
 C21H22O4
Molecular Weight:
 338.40
 Beilstein/REAXYS Number:
 4534154
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More