• USA Home
  • Search Results

  • lightgreensfyellowish79285514120811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Light Green SF Yellowish

3 Products
Synonym:   Acid Green 5
Image of Light Green SF Yellowish
CAS Number:
Linear Formula:
 C37H34N2O9S3Na2
Molecular Weight:
 792.85
 Beilstein/REAXYS Number:
 5717828
 Colour Index Number:
 42095
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Special Grade
  Solubility
Show More