• USA Home
  • Search Results

  • linalool154257870611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Linalool

7 Products
Synonym:   (±)-3,7-Dimethyl-1,6-octadien-3-ol, (±)-3,7-Dimethyl-3-hydroxy-1,6-octadiene
Image of Linalool
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2C(CH3)(OH)CH=CH2
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 1721488
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Solubility
  Potency
  Shelf Life
  Assay
  Special Grade
Show More