• USA Home
  • Search Results

  • linalylacetate1962911595711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Linalyl acetate

5 Products
Synonym:   3,7-Dimethyl-1,6-octadien-3-yl acetate, Bergamol
Image of Linalyl acetate
CAS Number:
Linear Formula:
 CH3CO2C(CH=CH2)(CH3)CH2CH2CH=C(CH3)2
Molecular Weight:
 196.29
 Beilstein/REAXYS Number:
 1724500
 Council of Europe no.:
 203
 EC Number:
 FEMA Number:
 2636
 Flavis number:
 9.013
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Shelf Life
  Allergen
  Organoleptic property
  Titertest
  Biological Source
  Assay
  Special Grade
Show More