• USA Home
  • Search Results

  • linalylformate1822611599111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Linalyl formate

1 Products
Image of Linalyl formate
CAS Number:
Linear Formula:
 HCO2C(C=CH2)(CH3)CH2CH2CH=C(CH3)2
Molecular Weight:
 182.26
 Council of Europe no.:
 347
 EC Number:
 FEMA Number:
 2642
 Flavis number:
 9.080
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More

Product #
Description
W264202 Linalyl formate, FCC,