• USA Home
  • Search Results

  • linarin5925548036411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Linarin

1 Products
Synonym:   5,7-Dihydroxy 4′-methoxyflavone 7-rutinoside, 7-((6-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl)oxy)-5-hydroxy-2-(4-methoxyphenyl)-4H-benzopyran-4-one, Acacetin-7-rutinoside, Buddleoside, Linarigenin glycoside
Image of Linarin
CAS Number:
Empirical Formula (Hill Notation):
 C28H32O14
Molecular Weight:
 592.55
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More