• USA Home
  • Search Results

  • lindane290835889911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lindane

4 Products
Synonym:   γ-BHC, 1α,2α,3β,4α,5α,6β-Hexachlorocyclohexane
Image of Lindane
CAS Number:
Empirical Formula (Hill Notation):
 C6H6Cl6
Molecular Weight:
 290.83
 Beilstein/REAXYS Number:
 1907337
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Solubility
  Special Grade
Show More