• USA Home
  • Search Results

  • linezolid3373516580003311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Linezolid

3 Products
Synonym:   N-[[(5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide, PNU-100766, U-100766
Image of Linezolid
CAS Number:
Empirical Formula (Hill Notation):
 C16H20FN3O4
Molecular Weight:
 337.35
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More