• USA Home
  • Search Results

  • linseedflaxseedoil12345800126111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Linseed (flaxseed) oil

1 Products
Synonym:   Linseed oil
CAS Number:
 EC Number:
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More