• USA Home
  • Search Results

  • lipasefromaspergillusoryzae12345900162111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lipase from Aspergillus oryzae

3 Products
Synonym:   Lipolase 100L
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.1.1.3.
 MDL number:
 NACRES:
 NA.54
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Specific Activity
  Physical Form
Show More