• USA Home
  • Search Results

  • lipasefrompseudomonascepacia12345900162111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lipase from Pseudomonas cepacia

1 Products
Synonym:   PS Lipase, Triacylglycerol acylhydrolase, Triacylglycerol lipase
CAS Number:
 EC Number:
 Enzyme Commission (EC) Number:
 3.1.1.3  ( BRENDA | IUBMB )
 MDL number:
 NACRES
 NA.54
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More