• USA Home
  • Search Results

  • lithiumacetatedihydrate10202610817411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium acetate dihydrate

3 Products
Synonym:   Acetic acid lithium salt
Image of Lithium acetate dihydrate
CAS Number:
Linear Formula:
 CH3COOLi · 2H2O
Molecular Weight:
 102.02
 Beilstein/REAXYS Number:
 3564320
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  pH Value
  UV-Absorption Values
  Solubility
  Product Line
Show More