• USA Home
  • Search Results

  • lithiumbenzoate1280555354811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium benzoate

1 Products
Synonym:   Benzoic acid lithium salt
Image of Lithium benzoate
CAS Number:
Linear Formula:
 C6H5COOLi
Molecular Weight:
 128.05
 Beilstein Registry Number:
 3598089
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
201162 Lithium benzoate, 99%,