• USA Home
  • Search Results

  • lithiumchloride4239744741811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium chloride

14 Products
Image of Lithium chloride
CAS Number:
Linear Formula:
 LiCl
Molecular Weight:
 42.39
 EC Number:
 MDL number:
 NACRES:
 NA.31
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Values
  Quality clarifier
  Potency
  Particle Dimension
  Melting Point
  Vapor pressure
  Loss
  Product Line
  pH Value
  Solubility
  Special Grade
  Physical Form
Show More

Product #
Description
L9650 Lithium chloride, for molecular biology, ≥99%,
203637 Lithium chloride, powder, ≥99.98% trace metals basis,
310468 Lithium chloride, ACS reagent, ≥99%,
L4408 Lithium chloride, BioXtra, ≥99.0% (titration),
793620 Lithium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%,
213233 Lithium chloride, ReagentPlus®, 99%,
746460 Lithium chloride, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%,
62476 Lithium chloride, BioUltra, for molecular biology, anhydrous, ≥99.0% (AT),
1.05679 Lithium chloride, for analysis EMSURE® ACS,Reag. Ph Eur,
429457 Lithium chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis,
916013 Lithium chloride, anhydrous, 99.95% trace metals basis,
449881 Lithium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis,
73036 Lithium chloride, puriss. p.a., anhydrous, ≥99.0% (AT),
LX0331 Lithium Chloride, Meets ACS Specifications GR ACS,