• USA Home
  • Search Results

  • lithiumchloridepotassiumchlorideeutectic123456556796611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium chloride/potassium chloride eutectic

1 Products
Synonym:   Lithium potassium chloride
Image of Lithium chloride/potassium chloride eutectic
CAS Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More