• USA Home
  • Search Results

  • lithiumdiisopropylamidesolution10712411154011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Lithium diisopropylamide solution

2 Products
Synonym:   LDA
Image of Lithium diisopropylamide solution
CAS Number:
Linear Formula:
 [(CH3)2CH]2NLi
Molecular Weight:
 107.12
 Beilstein/REAXYS Number:
 3655042
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More